Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MID2
Entrez Gene ID
851042
Gene Name
Gene Aliases
KAI1, YLR332W
Chromosome Location
Chr.XII: 790676 - 791806 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182221.1 NP_013436.1 1 - 1 1057 96

View Full Product Details