Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPO77
Entrez Gene ID
851054
Gene Name
Gene Aliases
YLR341W
Chromosome Location
Chr.XII: 807385 - 808818 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182230.1 NP_013445.1 1 - 1 1255 75

View Full Product Details