Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAC1
Entrez Gene ID
855035
Gene Name
Gene Aliases
CUA1, YMR021C
Chromosome Location
Chr.XIII: 317165 - 318418 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182517.1 NP_013734.1 1 - 1 1151 92

View Full Product Details