Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLA4
Entrez Gene ID
855418
Gene Name
Gene Aliases
ERC10, YNL298W
Chromosome Location
Chr.XIV: 68915 - 71443 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183136.1 NP_014101.1 1 - 1 2446 89

View Full Product Details