Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
URK1
Entrez Gene ID
855746
Gene Name
Gene Aliases
YNR012W
Chromosome Location
Chr.XIV: 647432 - 648937 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183189.1 NP_014409.1 1 - 1 1250 91

View Full Product Details