Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FSH3
Entrez Gene ID
854454
Gene Name
Gene Aliases
YOR280C
Chromosome Location
Chr.XV: 844992 - 845792 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183699.1 NP_014923.1 1 - 1 651 87

View Full Product Details