Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA11
Entrez Gene ID
855842
Gene Name
Gene Aliases
CLS9, TFP3, YPL234C
Chromosome Location
Chr.XVI: 104946 - 105440 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184048.1 NP_015090.1 1 - 1 355 98

View Full Product Details