Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QKI
Entrez Gene ID
492277
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
PqkI
Chromosome Location
Chr.1: 4732575 - 4883386 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007195.1 NP_001007196.1 5 - 6 633 61
XM_021083643.1 XP_020939302.1 - 778 61
XM_021083671.1 XP_020939330.1 - 768 61
XM_021083682.1 XP_020939341.1 - 1295 61
XM_021083688.1 XP_020939347.1 - 767 61
GenBank mRNA AB177988.1 - 5 - 6 633 61
AK390725.1 - 4 - 5 460 61

View Full Product Details