Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YWHAQ
Entrez Gene ID
100048964
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta
Gene Aliases
-
Chromosome Location
Chr.3: 126738795 - 126772784 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021087862.1 XP_020943521.1 - 716 129
GenBank mRNA AK234468.1 - 5 - 5 558 129
AK235440.1 - 3 - 3 421 129
AM503090.1 - 3 - 3 247 129

View Full Product Details