Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
733576
Gene Name
beclin 1
Gene Aliases
ATG6
Chromosome Location
Chr.12: 20041914 - 20054249 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044530.1 NP_001037995.1 12 - 12 1352 100
XM_013980932.2 XP_013836386.1 - 1292 100
GenBank mRNA AK235833.1 - 12 - 12 1347 100
AK390423.1 - 12 - 12 1361 100
AM051354.1 - 12 - 12 1352 100

View Full Product Details