Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN3
Entrez Gene ID
100049663
Gene Name
endothelin 3
Gene Aliases
-
Chromosome Location
Chr.17: 59418667 - 59448382 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098582.2 NP_001092052.1 4 - 5 609 89
XM_021077167.1 XP_020932826.1 - 89
GenBank mRNA AB271922.1 - 4 - 5 609 89
AK236444.1 - 4 - 5 89

View Full Product Details