Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF19A
Entrez Gene ID
654326
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
RFN19, RNF19
Chromosome Location
Chr.4: 36654197 - 36727514 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038006.1 NP_001033095.1 2 - 3 888 102
XM_005662947.3 XP_005663004.1 - 1513 102
XM_013996572.2 XP_013852026.1 - 1284 102
XM_013996573.2 XP_013852027.1 - 1372 102
XM_013996574.2 XP_013852028.1 - 1383 102
GenBank mRNA DQ091176.1 - 2 - 3 888 102

View Full Product Details