Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLF12
Entrez Gene ID
100174955
Gene Name
Kruppel like factor 12
Gene Aliases
-
Chromosome Location
Chr.11: 45981096 - 46655472 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134347.2 NP_001127819.1 6 - 7 1064 110
XM_021065113.1 XP_020920772.1 - 1289 110
XM_021065114.1 XP_020920773.1 - 1460 110
XM_021065115.1 XP_020920774.1 - 1502 110
XM_021065116.1 XP_020920775.1 - 1310 110
XM_021065117.1 XP_020920776.1 - 7267 110
XM_021065118.1 XP_020920777.1 - 1479 110
XM_021065119.1 XP_020920778.1 - 1524 110
XM_021065120.1 XP_020920779.1 - 1446 110
XM_021065121.1 XP_020920780.1 - 1381 110
XM_021065122.1 XP_020920781.1 - 1535 110
XM_021065123.1 XP_020920782.1 - 1469 110
XM_021065124.1 XP_020920783.1 - 1379 110
XM_021065125.1 XP_020920784.1 - 1225 110
XM_021065126.1 XP_020920785.1 - 1402 110
XM_021065127.1 XP_020920786.1 - 1450 110
GenBank mRNA AK232583.1 - 7 - 8 1330 110
AK233199.1 - 8 - 9 1330 110
AK235805.1 - 7 - 8 1090 110
AK237226.1 - 9 - 10 1479 110
AK238895.1 - 7 - 8 1344 110
AK395759.1 - 7 - 8 1232 110
EU669079.2 - 6 - 7 1064 110

View Full Product Details