Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMC4
Entrez Gene ID
100153736
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
TMEM85
Chromosome Location
Chr.7: 80266001 - 80270691 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243441.1 NP_001230370.1 2 - 3 316 64
GenBank mRNA AK231739.1 - 2 - 3 316 64
AK348629.1 - 2 - 3 265 64
AK397511.1 - 2 - 3 311 64
AY609894.1 - 2 - 3 294 64

View Full Product Details