Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMG1
Entrez Gene ID
100627558
Gene Name
EMG1 N1-specific pseudouridine methyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 63746326 - 63751469 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243478.1 NP_001230407.1 3 - 4 506 68
GenBank mRNA AK231488.1 - 3 - 4 508 68
AK240399.1 - 3 - 4 425 68
AK346690.1 - 3 - 4 425 68
AY610201.1 - 3 - 4 482 68

View Full Product Details