Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANR
Entrez Gene ID
100232981
Gene Name
anthocyanidin reductase
Gene Aliases
GSVIVT00005344001, ban
Chromosome Location
Chr.Un_NW_002240272: 13945 - 17265 on Build 12X
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DQ129684.1 - 1 - 2 132 70

View Full Product Details