Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
snrpd1
Entrez Gene ID
447999
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
-
Chromosome Location
Chr.6: 82271946 - 82278628 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004780.1 NP_001004780.1 3 - 4 723 147
XM_012964505.2 XP_012819959.1 3 - 4 724 147
GenBank mRNA BC074514.1 - 3 - 4 723 147
CT025480.2 - 3 - 4 307 147

View Full Product Details