Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp2r1a
Entrez Gene ID
395042
Gene Name
protein phosphatase 2 regulatory subunit A, alpha
Gene Aliases
pp2a-aalpha, pp2aaalpha, ppp2r1a-a, ppp2r1a-b, pr65a
Chromosome Location
Chr.7: 111894772 - 111920217 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204074.1 NP_989405.1 7 - 8 924 59
GenBank mRNA BC064863.1 - 7 - 8 924 59
CR848503.2 - 7 - 8 992 59

Gene Symbol
LOC101731355
Entrez Gene ID
101731355
Gene Name
serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alpha isoform-like
Gene Aliases
-
Chromosome Location
Chr.7: 111954203 - 111962222 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004920454.3 XP_004920511.2 6 - 7 904 59

View Full Product Details