Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wbp11
Entrez Gene ID
448171
Gene Name
WW domain binding protein 11
Gene Aliases
-
Chromosome Location
Chr.4: 117899956 - 117913259 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005672.1 NP_001005672.1 3 - 4 228 80
GenBank mRNA BC074710.1 - 3 - 4 228 80
CR848543.2 - 3 - 4 189 80

View Full Product Details