Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tub
Entrez Gene ID
493219
Gene Name
tubby bipartite transcription factor
Gene Aliases
rd5, tubby
Chromosome Location
Chr.Un|NW_016683977: 2277 - 5054 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007492.1 NP_001007493.1 - 495 64
GenBank mRNA BC079929.1 - - 495 64

View Full Product Details