Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnf14
Entrez Gene ID
493480
Gene Name
ring finger protein 14
Gene Aliases
ara54, hfb30, hrihfb2038, triad2
Chromosome Location
Chr.3: 28120299 - 28141792 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008117.1 NP_001008118.1 5 - 6 1303 76
GenBank mRNA BC081322.1 - 5 - 6 1303 76

Gene Symbol
LOC101732063
Entrez Gene ID
101732063
Gene Name
E3 ubiquitin-protein ligase RNF14-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683854: 33365 - 48107 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012954767.2 XP_012810221.1 4 - 5 1037 76

View Full Product Details