Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
emc10
Entrez Gene ID
394497
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
c19orf63
Chromosome Location
Chr.7: 102570268 - 102577736 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203571.1 NP_988902.1 5 - 6 591 99
XM_012956621.2 XP_012812075.1 5 - 6 678 99
GenBank mRNA BC061625.1 - 5 - 6 591 99
CR760224.2 - 5 - 6 596 99
CR760298.2 - 1 - 2 76 99

View Full Product Details