Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp23
Entrez Gene ID
594990
Gene Name
ATP23 metallopeptidase and ATP synthase assembly factor homolog
Gene Aliases
xrcc6bp1
Chromosome Location
Chr.3: 53799923 - 53807336 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030431.1 NP_001025602.1 3 - 4 319 89
XM_018092119.1 XP_017947608.1 2 - 3 158 89
GenBank mRNA BC091052.1 - 3 - 4 319 89
CR761179.2 - 3 - 4 294 89

View Full Product Details