Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
lsm14b
Entrez Gene ID
595068
Gene Name
LSM family member 14B
Gene Aliases
ba11m20.3, fam61b, ft005, lsm13, rap42, rap46
Chromosome Location
Chr.10: 1122608 - 1132385 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030505.1 NP_001025676.1 3 - 4 466 76
GenBank mRNA BC093463.1 - 3 - 4 466 76

View Full Product Details