Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab19
Entrez Gene ID
550000
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
rab19b
Chromosome Location
Chr.3: 208919 - 213070 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017246.2 NP_001017246.1 2 - 3 321 99
XM_012959096.2 XP_012814550.1 2 - 3 532 99
GenBank mRNA CR760145.2 - 2 - 3 321 99

View Full Product Details