Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sssca1
Entrez Gene ID
550002
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
-
Chromosome Location
Chr.4: 42800258 - 42804431 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017248.2 NP_001017248.1 3 - 4 267 85
XM_012961190.2 XP_012816644.1 3 - 4 353 85
GenBank mRNA BC158901.1 - 3 - 4 287 85
CR760119.2 - 3 - 4 267 85

View Full Product Details