Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cep44
Entrez Gene ID
780008
Gene Name
centrosomal protein 44kDa
Gene Aliases
kiaa1712, ps1tp3
Chromosome Location
Chr.1: 39128042 - 39142296 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079085.1 NP_001072553.1 8 - 9 1179 75
XM_012965471.2 XP_012820925.1 8 - 9 1092 75
XM_018094499.1 XP_017949988.1 7 - 8 1038 75
GenBank mRNA BC122069.1 - 8 - 9 1179 75
CR760216.2 - 8 - 9 1223 75

View Full Product Details