Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rtkn2
Entrez Gene ID
100145618
Gene Name
rhotekin 2
Gene Aliases
-
Chromosome Location
Chr.7: 9730399 - 9789780 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127024.1 NP_001120496.1 2 - 3 270 65
XM_012966095.2 XP_012821549.1 2 - 3 445 65
GenBank mRNA BC161376.1 - 2 - 3 270 65
BC161383.1 - 2 - 3 270 65
CR942636.2 - 2 - 3 327 65

View Full Product Details