Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dapk2
Entrez Gene ID
100145135
Gene Name
death-associated protein kinase 2
Gene Aliases
drp-1, drp1
Chromosome Location
Chr.3: 81554522 - 81604617 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126642.1 NP_001120114.1 10 - 11 1019 73
XM_012966054.2 XP_012821508.1 10 - 11 1096 73
GenBank mRNA BC158982.1 - 10 - 11 1019 73

View Full Product Details