Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmcc1
Entrez Gene ID
100170168
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
-
Chromosome Location
Chr.4: 121603287 - 121687392 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142914.1 NP_001136386.1 5 - 6 1189 81
XM_012960654.2 XP_012816108.1 5 - 6 1261 81
XM_012960655.2 XP_012816109.1 4 - 5 1169 81
XM_012960656.1 XP_012816110.1 2 - 3 408 81
XM_012960657.2 XP_012816111.1 2 - 3 370 81
XM_012960658.2 XP_012816112.1 2 - 3 496 81
XM_012960659.1 XP_012816113.1 2 - 3 518 81
XM_012960660.2 XP_012816114.1 2 - 3 590 81
XM_012960662.2 XP_012816116.1 2 - 3 433 81
XM_018092939.1 XP_017948428.1 6 - 7 1414 81
GenBank mRNA BC167584.1 - 5 - 6 1189 81
CT025256.2 - 4 - 5 1156 81

View Full Product Details