Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
IPT5
Entrez Gene ID
100158226
Gene Name
isopentenyl transferase IPT5
Gene Aliases
ZEAMMB73_194180
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127722.1 NP_001121194.1 - 951 81

View Full Product Details