Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
kan3
Entrez Gene ID
100279550
Gene Name
kanadi3
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001152547.1 NP_001146019.1 - 1051 88
GenBank mRNA BT054439.1 - - 1051 88

View Full Product Details