Gene Transcripts

Gene Symbol
Vmn2r29
Entrez Gene ID
76229
Gene Name
vomeronasal 2, receptor 29
Gene Aliases
6430701C03Rik
Chromosome Location
Chr. 7: 7212996 - 7278289 on Build GRCm38
UniGene ID
Mm.387947
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003555.1 7 1141
GenBank mRNA AK018294.1 - 7 1079
AK039785.1 - 1 3004
BC096375.1 - 7 1067

View Full Product Details