Gene Transcripts

Gene Symbol
Rbmx
Entrez Gene ID
19655
Gene Name
RNA binding motif protein, X chromosome
Gene Aliases
hnRNP G
Chromosome Location
Chr. X: 57383348 - 57393042 on Build GRCm38
UniGene ID
Mm.28275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_029425.1 10 2233
GenBank mRNA AJ237846.1 - 10 2163
AK004162.1 - 1 309
AK087940.1 - 4 1338

View Full Product Details