Gene Transcripts

Gene Symbol
TATDN1
Entrez Gene ID
83940
Gene Name
TatD DNase domain containing 1
Gene Aliases
CDA11
Chromosome Location
Chr. 8: 124488494 - 124539088 on Build GRCh38
UniGene ID
Hs.170568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317889.1 NP_001304818.1 10 722
NR_027427.1 10 726
XM_006716666.3 XP_006716729.1 10 742
XM_006716667.3 XP_006716730.1 10 787
XM_006716668.3 XP_006716731.1 10 800
XM_006716669.3 XP_006716732.1 8 684
XM_006716670.3 XP_006716733.1 9 643
XM_011517331.2 XP_011515633.1 9 638
GenBank mRNA BX332564.2 - 10 704

View Full Product Details