Gene Transcripts
- Gene Symbol
- FAM172A
- Entrez Gene ID
- 83989
- Gene Name
- family with sequence similarity 172 member A
- Gene Aliases
- C5orf21
- Chromosome Location
- Chr. 5: 93615130 - 94111699 on Build GRCh38
- UniGene ID
- Hs.600086
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028080.1 |
|
3, 4 |
416 |
|
XM_005272108.3 |
XP_005272165.1
|
3, 4 |
411 |
|
XM_006714718.3 |
XP_006714781.1
|
2, 3 |
1123 |
|
XM_006714719.3 |
XP_006714782.1
|
2, 3 |
272 |
|
XM_011543673.2 |
XP_011541975.1
|
3, 4 |
312 |
|
XM_017009957.1 |
XP_016865446.1
|
2, 3 |
489 |
|
XM_017009958.1 |
XP_016865447.1
|
2, 3 |
284 |
|
XM_017009960.1 |
XP_016865449.1
|
2, 3 |
258 |
|
GenBank mRNA |
AK295127.1 |
- |
3, 4 |
416 |
|
AK297471.1 |
- |
2, 3 |
178 |
|
DC324758.1 |
- |
3, 4 |
416 |
|
DC374283.1 |
- |
3, 4 |
401 |
|
View Full Product Details