Gene Transcripts

Gene Symbol
PRNT
Entrez Gene ID
149830
Gene Name
prion protein (testis specific)
Gene Aliases
M8
Chromosome Location
Chr. 20: 4731282 - 4740668 on Build GRCh38
UniGene ID
Hs.126516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024267.1 2 1053
NR_024268.1 2 816
NR_024269.1 2 593
GenBank mRNA AJ427539.1 - 2 817
AJ427540.1 - 2 1053
AL137296.1 - 2 593
BC137389.1 - 1 470
BC137390.1 - 1 471

View Full Product Details