Gene Transcripts

Gene Symbol
SARNP
Entrez Gene ID
84324
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
CIP29, HCC1, HSPC316, THO1
Chromosome Location
Chr. 12: 55752463 - 55817756 on Build GRCh38
UniGene ID
Hs.505676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026722.1 11 787
NR_026723.1 12 834
GenBank mRNA AK290508.1 - 12 802
BC093051.1 - 11 753 30382893


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB636681.1 - 5 466 Chr. 12 - 55752579 - 55789146 [-]
GenBankmRNA BG209498.1 - 3 283 Chr. 12 - 55752577 - 55760642 [-]
GenBankmRNA BG204693.1 - 3 285 Chr. 12 - 55752577 - 55760638 [-]
GenBankmRNA BE885590.1 - 8 543 Chr. 12 - 55752627 - 55796076 [-]
GenBankmRNA BG188761.1 - 3 278 Chr. 12 - 55752577 - 55760618 [-]
GenBankmRNA BM688973.1 - 4 355 Chr. 12 - 55752695 - 55789144 [-]
GenBankmRNA BG210608.1 - 3 284 Chr. 12 - 55752577 - 55760642 [-]
GenBankmRNA BM797801.1 - 5 367 Chr. 12 - 55752688 - 55790586 [-]
GenBankmRNA BG192927.1 - 3 282 Chr. 12 - 55752577 - 55760642 [-]
GenBankmRNA AA429710.1 - 5 364 Chr. 12 - 55752690 - 55790583 [-]
GenBankmRNA BG184024.1 - 3 281 Chr. 12 - 55752577 - 55760642 [-]
GenBankmRNA AA491996.1 - 2 63 Chr. 12 - 55752577 - 55757421 [-]
GenBankmRNA DB524653.1 - 2 164 Chr. 12 - 55752583 - 55757525 [-]
GenBankmRNA BG196491.1 - 3 280 Chr. 12 - 55752577 - 55760642 [-]
GenBankmRNA AW993022.1 - 7 428 Chr. 12 - 55752757 - 55794832 [-]
GenBankmRNA BG214717.1 - 3 283 Chr. 12 - 55752577 - 55760642 [-]

View Full Product Details