Gene Transcripts
- Gene Symbol
- MORC2-AS1
- Entrez Gene ID
- 150291
- Gene Name
- MORC2 antisense RNA 1
- Gene Aliases
- C22orf27, NCRNA00325
- Chromosome Location
- Chr. 22: 30922308 - 30926654 on Build GRCh38
- UniGene ID
- Hs.592202
- Species
- Homo sapiens
- Gene Symbol
- MORC2
- Entrez Gene ID
- 22880
- Gene Name
- MORC family CW-type zinc finger 2
- Gene Aliases
- CMT2Z, ZCW3, ZCWCC1
- Chromosome Location
- Chr. 22: 30925130 - 30968497 on Build GRCh38
- UniGene ID
- Hs.555918
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM683680.1 |
- |
1 |
91 |
|
View Full Product Details