Gene Transcripts

Gene Symbol
MORC2-AS1
Entrez Gene ID
150291
Gene Name
MORC2 antisense RNA 1
Gene Aliases
C22orf27, NCRNA00325
Chromosome Location
Chr. 22: 30922308 - 30926654 on Build GRCh38
UniGene ID
Hs.592202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026920.1 2 1394
GenBank mRNA AK093120.1 - 2 1293
AK098685.1 - 1 775
BC042980.1 - 3 1533
BC065553.1 - 2 1287


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000422995.2 - 3 1534 Chr. 22 - 30922345 - 30926654 [+]
GENCODE ENST00000432624.2 - 2 1394 Chr. 22 - 30922308 - 30926550 [+]

View Full Product Details