Gene Transcripts
- Gene Symbol
- MORC2-AS1
- Entrez Gene ID
- 150291
- Gene Name
- MORC2 antisense RNA 1
- Gene Aliases
- C22orf27, NCRNA00325
- Chromosome Location
- Chr. 22: 30922308 - 30926654 on Build GRCh38
- UniGene ID
- Hs.592202
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000422995.2 |
- |
3 |
698 |
|
Chr. 22 - 30922345 - 30926654 [+] |
GENCODE |
ENST00000432624.2 |
- |
2 |
558 |
|
Chr. 22 - 30922308 - 30926550 [+] |
View Full Product Details