Gene Transcripts
- Gene Symbol
- MUM1
- Entrez Gene ID
- 84939
- Gene Name
- melanoma associated antigen (mutated) 1
- Gene Aliases
- EXPAND1, HSPC211, MUM-1
- Chromosome Location
- Chr. 19: 1354977 - 1378431 on Build GRCh38
- UniGene ID
- Hs.515016
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY063060.1 |
- |
2, 3 |
197 |
|
Chr. 19 - 1358378 - 1360461 [+] |
GenBankmRNA |
DC335695.1 |
- |
3, 4 |
212 |
|
Chr. 19 - 1357064 - 1360496 [+] |
GenBankmRNA |
CN430689.1 |
- |
4, 5 |
409 |
|
Chr. 19 - 1356323 - 1360478 [+] |
GenBankmRNA |
DC410184.1 |
- |
5, 6 |
427 |
|
Chr. 19 - 1354988 - 1360294 [+] |
GenBankmRNA |
DR158298.1 |
- |
5, 6 |
415 |
|
Chr. 19 - 1354999 - 1360452 [+] |
View Full Product Details