Gene Transcripts
- Gene Symbol
- SNHG12
- Entrez Gene ID
- 85028
- Gene Name
- small nucleolar RNA host gene 12
- Gene Aliases
- ASLNC04080, C1orf79, LINC00100, NCRNA00100
- Chromosome Location
- Chr. 1: 28578538 - 28581854 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE794361.1 |
- |
6 |
688 |
|
Chr. 1 - 28578538 - 28581834 [-] |
GenBankmRNA |
BM747774.1 |
- |
1 |
345 |
|
Chr. 1 - 28578538 - 28578912 [-] |
GenBankmRNA |
EB387922.1 |
- |
4 |
330 |
|
Chr. 1 - 28578536 - 28580649 [-] |
View Full Product Details