Gene Transcripts
- Gene Symbol
- SNHG12
- Entrez Gene ID
- 85028
- Gene Name
- small nucleolar RNA host gene 12
- Gene Aliases
- ASLNC04080, C1orf79, LINC00100, NCRNA00100
- Chromosome Location
- Chr. 1: 28578538 - 28581854 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN260333.1 |
- |
5 |
575 |
|
Chr. 1 - 28578597 - 28581859 [-] |
GenBankmRNA |
BE794361.1 |
- |
5 |
550 |
|
Chr. 1 - 28578538 - 28581834 [-] |
GenBankmRNA |
BF765335.1 |
- |
4 |
260 |
|
Chr. 1 - 28578636 - 28581171 [-] |
GenBankmRNA |
AV697366.1 |
- |
4 |
519 |
|
Chr. 1 - 28579532 - 28581040 [-] |
GenBankmRNA |
EB387922.1 |
- |
3 |
192 |
|
Chr. 1 - 28578536 - 28580649 [-] |
View Full Product Details