Gene Transcripts

Gene Symbol
TRIM78P
Entrez Gene ID
117852
Gene Name
tripartite motif containing 78, pseudogene
Gene Aliases
TRIMP1
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.742023
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF230412.1 - LIT2077
TRIMP pseudogene
2 381
AL360159.1 -
2 381
AL360183.1 -
1 129

View Full Product Details