Gene Transcripts

Gene Symbol
SNORA21
Entrez Gene ID
619505
Gene Name
small nucleolar RNA, H/ACA box 21
Gene Aliases
ACA21
Chromosome Location
Chr. 17: 38852863 - 38852995 on Build GRCh38
UniGene ID
Hs.738742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002576.1 1 107
GenBank mRNA AJ609436.1 - 1 107
AK095288.1 - 1 854


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA504097.1 - 2 247 Chr. 17 - 38852604 - 38853727 [-]
GenBankmRNA BM675662.1 - 2 255 Chr. 17 - 38852723 - 38853735 [-]
GenBankmRNA DA435180.1 - 1 418 Chr. 17 - 38852736 - 38853306 [-]
GenBankmRNA HY053696.1 - 1 444 Chr. 17 - 38852846 - 38853332 [-]
GenBankmRNA DB108830.1 - 2 256 Chr. 17 - 38852604 - 38853736 [-]
GenBankmRNA DA347435.1 - 1 124 Chr. 17 - 38852443 - 38853012 [-]

View Full Product Details