Gene Transcripts
- Gene Symbol
- SCARNA17
- Entrez Gene ID
- 677769
- Gene Name
- small Cajal body-specific RNA 17
- Gene Aliases
- U91
- Chromosome Location
- Chr. 18: 49814023 - 49814443 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003003.2 |
|
1 |
37 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CB997907.1 |
- |
1 |
64 |
|
Chr. 18 - 49814023 - 49814677 [+] |
View Full Product Details