Gene Transcripts
- Gene Symbol
- SCARNA6
- Entrez Gene ID
- 677772
- Gene Name
- small Cajal body-specific RNA 6
- Gene Aliases
- U88
- Chromosome Location
- Chr. 2: 233288676 - 233288941 on Build GRCh38
- UniGene ID
- Hs.689637
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003006.1 |
|
1 |
86 |
|
GenBank mRNA |
AY077738.1 |
- |
1 |
86 |
|
View Full Product Details