Gene Transcripts

Gene Symbol
SNORA29
Entrez Gene ID
677812
Gene Name
small nucleolar RNA, H/ACA box 29
Gene Aliases
ACA29
Chromosome Location
Chr. 6: 159785594 - 159785733 on Build GRCh38
UniGene ID
Hs.363137
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002965.1 1 26
GenBank mRNA AJ609472.1 - 1 26


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA429995.1 - 1 164 Chr. 6 - 159784784 - 159785871 [-]

View Full Product Details