Gene Transcripts
- Gene Symbol
- AQP7P3
- Entrez Gene ID
- 441432
- Gene Name
- aquaporin 7 pseudogene 3
- Gene Aliases
- AQPap-3
- Chromosome Location
- Chr. 9: 65877574 - 65912560 on Build GRCh38
- UniGene ID
- Hs.743215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026558.1 |
|
2, 3 |
119 |
|
GenBank mRNA |
BC052332.1 |
- |
2, 3 |
119 |
|
View Full Product Details